Assay Details
| Gene Symbol: LOC100340255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347053.1 | XP_017202542.1 | 10 - 11 | 4408 | 101 | |
| XM_017347054.1 | XP_017202543.1 | 9 - 10 | 3515 | 101 | ||
More Information
Set Membership: |
|

LOC100340255
Genomic Map