Assay Details
| Gene Symbol: LOC100340255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347053.1 | XP_017202542.1 | 11 - 12 | 4468 | 88 | |
| XM_017347054.1 | XP_017202543.1 | 10 - 11 | 3575 | 88 | ||
More Information
Set Membership: |
|

LOC100340255
Genomic Map