Assay Details
| Gene Symbol: LOC100340255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347053.1 | XP_017202542.1 | 13 - 14 | 4847 | 68 | |
| XM_017347054.1 | XP_017202543.1 | 12 - 13 | 3954 | 68 | ||
More Information
Set Membership: |
|

LOC100340255
Genomic Map