Assay Details
| Gene Symbol: NUAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717563.3 | XP_002717609.1 | 2 - 3 | 495 | 74 | |
| XM_008268520.2 | XP_008266742.1 | 2 - 3 | 263 | 74 | ||
| XM_008268521.2 | XP_008266743.1 | 2 - 3 | 428 | 74 | ||
| XM_008268522.2 | XP_008266744.1 | 2 - 3 | 428 | 74 | ||
More Information
Set Membership: |
|

NUAK2
Genomic Map