Assay Details
| Gene Symbol: IFNLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716020.2 | XP_002716066.1 | 6 - 7 | 1007 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFNLR1| Gene Symbol: IFNLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716020.2 | XP_002716066.1 | 6 - 7 | 1007 | 65 | |
Set Membership: |
|