Assay Details
| Gene Symbol: HFE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714212.3 | XP_002714258.1 | 4 - 5 | 1328 | 69 | |
| XM_008262463.2 | XP_008260685.1 | 4 - 5 | 1352 | 69 | ||
More Information
Set Membership: |
|

HFE
Genomic Map