Assay Details
Gene Symbol: C1H11orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708449.3 | XP_002708495.1 | 1 - 2 | 150 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002708449.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
