Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248745.2 | XP_008246967.1 | 493 | 84 | ||
| XM_008248746.1 | XP_008246968.1 | 532 | 84 | |||
| XM_017337758.1 | XP_017193247.1 | 635 | 84 | |||
More Information
Set Membership: |
|

CD2BP2
Genomic Map