Assay Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718378.3 | XP_002718424.2 | 6 - 7 | 2571 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A12| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718378.3 | XP_002718424.2 | 6 - 7 | 2571 | 71 | |
Set Membership: |
|