Assay Details
| Gene Symbol: FAM126B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258951.2 | XP_008257173.1 | 7 - 8 | 658 | 70 | |
| XM_008258952.2 | XP_008257174.1 | 4 - 5 | 407 | 70 | ||
| XM_017343020.1 | XP_017198509.1 | 8 - 9 | 731 | 70 | ||
| XM_017343021.1 | XP_017198510.1 | 8 - 9 | 800 | 70 | ||
| XM_017343022.1 | XP_017198511.1 | 8 - 9 | 727 | 70 | ||
| XM_017343023.1 | XP_017198512.1 | 8 - 9 | 796 | 70 | ||
| XM_017343024.1 | XP_017198513.1 | 7 - 8 | 727 | 70 | ||
| XM_017343025.1 | XP_017198514.1 | 8 - 9 | 730 | 70 | ||
More Information
Set Membership: |
|

FAM126B
Genomic Map