Assay Details
| Gene Symbol: ZNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251914.2 | XP_008250136.1 | 256 | 64 | ||
| XM_008251915.2 | XP_008250137.1 | 443 | 64 | |||
| XM_008251917.2 | XP_008250139.1 | 223 | 64 | |||
| XM_008251918.2 | XP_008250140.1 | 410 | 64 | |||
More Information
Set Membership: |
|

ZNF7
Genomic Map