Assay Details
| Gene Symbol: COG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711663.2 | XP_002711709.2 | 3 - 4 | 1442 | 71 | |
| XM_017342324.1 | XP_017197813.1 | 3 - 4 | 1442 | 71 | ||
More Information
Set Membership: |
|

COG8
Genomic Map