Assay Details
| Gene Symbol: TBC1D20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710889.3 | XP_002710935.1 | 2 - 3 | 403 | 77 | |
| XM_008256178.2 | XP_008254400.1 | 3 - 4 | 475 | 77 | ||
| XM_017341712.1 | XP_017197201.1 | 2 - 3 | 392 | 77 | ||
| XM_017341713.1 | XP_017197202.1 | 1 - 2 | 139 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017341713.1 transcripts.
Set Membership: |
|

TBC1D20
Genomic Map