Assay Details
| Gene Symbol: LOC100351973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723571.3 | XP_002723617.1 | 115 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002723571.3 transcripts.
Set Membership: |
|

LOC100351973
Genomic Map