Assay Details
| Gene Symbol: KDM5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720016.3 | XP_002720062.1 | 16 - 17 | 2368 | 56 | |
| XM_008272647.2 | XP_008270869.1 | 16 - 17 | 2368 | 56 | ||
| XM_008272648.2 | XP_008270870.1 | 16 - 17 | 2365 | 56 | ||
| XM_017349757.1 | XP_017205246.1 | 15 - 16 | 2774 | 56 | ||
| XM_017349758.1 | XP_017205247.1 | 15 - 16 | 2245 | 56 | ||
| XM_017349759.1 | XP_017205248.1 | 16 - 17 | 2897 | 56 | ||
More Information
Set Membership: |
|

KDM5C
Genomic Map