Assay Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710359.3 | XP_002710405.1 | 5 - 6 | 821 | 65 | |
| XM_002710360.3 | XP_002710406.1 | 5 - 6 | 825 | 65 | ||
More Information
Set Membership: |
|

MAT2B
Genomic Map