Assay Details
| Gene Symbol: MRPS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721108.3 | XP_002721154.3 | 630 | 59 | ||
| XM_008274529.2 | XP_008272751.1 | 770 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPS27| Gene Symbol: MRPS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721108.3 | XP_002721154.3 | 630 | 59 | ||
| XM_008274529.2 | XP_008272751.1 | 770 | 59 | |||
Set Membership: |
|