Assay Details
| Gene Symbol: CDC14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342238.1 | XP_017197727.1 | 7 - 8 | 903 | 70 | |
| XM_017342239.1 | XP_017197728.1 | 7 - 8 | 903 | 70 | ||
| XM_017342245.1 | XP_017197734.1 | 7 - 8 | 903 | 70 | ||
| XM_017342252.1 | XP_017197741.1 | 7 - 8 | 903 | 70 | ||
| XM_017342254.1 | XP_017197743.1 | 7 - 8 | 903 | 70 | ||
| XM_017342258.1 | XP_017197747.1 | 7 - 8 | 903 | 70 | ||
| XM_017342261.1 | XP_017197750.1 | 903 | 70 | |||
More Information
Set Membership: |
|

CDC14B
Genomic Map