Assay Details
| Gene Symbol: RPP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718504.3 | XP_002718550.3 | 7 - 8 | 605 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPP30| Gene Symbol: RPP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718504.3 | XP_002718550.3 | 7 - 8 | 605 | 67 | |
Set Membership: |
|