Assay Details
| Gene Symbol: ARL14EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709028.3 | XP_002709074.1 | 2 - 3 | 596 | 107 | |
| XM_008269247.2 | XP_008267469.1 | 2 - 3 | 596 | 107 | ||
More Information
Set Membership: |
|

ARL14EP
Genomic Map