Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269507.2 | XP_008267729.1 | 9 - 10 | 870 | 82 | |
| XM_017348266.1 | XP_017203755.1 | 9 - 10 | 883 | 82 | ||
| XM_017348267.1 | XP_017203756.1 | 8 - 9 | 762 | 82 | ||
More Information
Set Membership: |
|

SLC25A21
Genomic Map