Assay Details
| Gene Symbol: LOC100350735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711435.3 | XP_002711481.1 | 2 - 3 | 237 | 77 | |
| XM_017342109.1 | XP_017197598.1 | 2 - 3 | 240 | 77 | ||
| XM_017342110.1 | XP_017197599.1 | 2 - 3 | 197 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002711435.3, XM_017342109.1, XM_017342110.1 transcripts.
Set Membership: |
|

LOC100350735
Genomic Map