Assay Details
| Gene Symbol: ROPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344650.1 | XP_017200139.1 | 4 - 5 | 458 | 56 | |
| XM_017344651.1 | XP_017200140.1 | 4 - 5 | 505 | 56 | ||
More Information
Set Membership: |
|

ROPN1L
Genomic Map