Assay Details
Gene Symbol: LOC100344961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017339502.1 | XP_017194991.1 | 2982 | 86 | ||
XM_017339503.1 | XP_017194992.1 | 2970 | 86 | |||
XM_017339505.1 | XP_017194994.1 | 2811 | 86 |

More Information
Set Membership: |
![]() ![]() |
