Assay Details
Gene Symbol: LOC100344961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008251850.1 | XP_008250072.1 | 2294 | 64 | ||
XM_017339500.1 | XP_017194989.1 | 2294 | 64 | |||
XM_017339501.1 | XP_017194990.1 | 2282 | 64 | |||
XM_017339502.1 | XP_017194991.1 | 2294 | 64 | |||
XM_017339503.1 | XP_017194992.1 | 2282 | 64 | |||
XM_017339504.1 | XP_017194993.1 | 2294 | 64 | |||
XM_017339505.1 | XP_017194994.1 | 2282 | 64 | |||
XM_017339506.1 | XP_017194995.1 | 2294 | 64 |

More Information
Set Membership: |
![]() ![]() |
