Assay Details
Gene Symbol: LOC100344961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017339500.1 | XP_017194989.1 | 2661 | 61 | ||
XM_017339501.1 | XP_017194990.1 | 2649 | 61 | |||
XM_017339502.1 | XP_017194991.1 | 2661 | 61 | |||
XM_017339503.1 | XP_017194992.1 | 2649 | 61 | |||
XM_017339504.1 | XP_017194993.1 | 2661 | 61 |

More Information
Set Membership: |
![]() ![]() |
