Assay Details
| Gene Symbol: LOC100344961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339500.1 | XP_017194989.1 | 2824 | 97 | ||
| XM_017339501.1 | XP_017194990.1 | 2812 | 97 | |||
| XM_017339502.1 | XP_017194991.1 | 2824 | 97 | |||
| XM_017339503.1 | XP_017194992.1 | 2812 | 97 | |||
More Information
Set Membership: |
|

LOC100344961
Genomic Map