Assay Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711391.3 | XP_002711437.1 | 4 - 5 | 458 | 79 | |
| XM_017342092.1 | XP_017197581.1 | 5 - 6 | 660 | 79 | ||
| XM_017342093.1 | XP_017197582.1 | 4 - 5 | 509 | 79 | ||
More Information
Set Membership: |
|

CBY1
Genomic Map