Assay Details
| Gene Symbol: LOC100351437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348165.1 | XP_017203654.1 | 1 - 2 | 186 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017348165.1 transcripts.
Set Membership: |
|

LOC100351437
Genomic Map