Assay Details
| Gene Symbol: LOC100351437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348165.1 | XP_017203654.1 | 39 - 40 | 5795 | 113 | |
| XM_017348166.1 | XP_017203655.1 | 39 - 40 | 5737 | 113 | ||
More Information
Set Membership: |
|

LOC100351437
Genomic Map