Assay Details
| Gene Symbol: LOC100351437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348165.1 | XP_017203654.1 | 9 - 10 | 1603 | 75 | |
| XM_017348166.1 | XP_017203655.1 | 9 - 10 | 1545 | 75 | ||
More Information
Set Membership: |
|

LOC100351437
Genomic Map