Assay Details
| Gene Symbol: PDILT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711767.3 | XP_002711813.1 | 9 - 10 | 1460 | 66 | |
| XM_017342475.1 | XP_017197964.1 | 9 - 10 | 1430 | 66 | ||
| XM_017342476.1 | XP_017197965.1 | 8 - 9 | 1293 | 66 | ||
| XM_017342477.1 | XP_017197966.1 | 8 - 9 | 1221 | 66 | ||
More Information
Set Membership: |
|

PDILT
Genomic Map