Assay Details
| Gene Symbol: RHOBTB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261963.2 | XP_008260185.1 | 11 - 12 | 1841 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RHOBTB3| Gene Symbol: RHOBTB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261963.2 | XP_008260185.1 | 11 - 12 | 1841 | 68 | |
Set Membership: |
|