Assay Details
| Gene Symbol: SLC35B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262360.2 | XP_008260582.1 | 4 - 5 | 665 | 82 | |
| XM_008262362.2 | XP_008260584.1 | 4 - 5 | 743 | 82 | ||
| XM_008262363.2 | XP_008260585.1 | 4 - 5 | 690 | 82 | ||
| XM_008262365.2 | XP_008260587.1 | 4 - 5 | 687 | 82 | ||
More Information
Set Membership: |
|

SLC35B3
Genomic Map