Assay Details
| Gene Symbol: MUC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268996.2 | XP_008267218.1 | 2 - 3 | 1088 | 92 | |
| XM_017345610.1 | XP_017201099.1 | 2 - 3 | 1104 | 92 | ||
More Information
Set Membership: |
|

MUC15
Genomic Map