Assay Details
| Gene Symbol: ZNF76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262812.2 | XP_008261034.1 | 7 - 8 | 824 | 56 | |
| XM_008262814.2 | XP_008261036.1 | 7 - 8 | 824 | 56 | ||
| XM_008262815.2 | XP_008261037.1 | 7 - 8 | 824 | 56 | ||
| XM_017345064.1 | XP_017200553.1 | 7 - 8 | 824 | 56 | ||
More Information
Set Membership: |
|

ZNF76
Genomic Map