Assay Details
| Gene Symbol: LOC100345100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269377.2 | XP_008267599.1 | 3 - 4 | 639 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100345100| Gene Symbol: LOC100345100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269377.2 | XP_008267599.1 | 3 - 4 | 639 | 90 | |
Set Membership: |
|