Assay Details
| Gene Symbol: LOC100352550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248916.2 | XP_008247138.1 | 1890 | 63 | ||
| XM_008248917.2 | XP_008247139.1 | 1738 | 63 | |||
More Information
Set Membership: |
|

LOC100352550
Genomic Map