Assay Details
| Gene Symbol: BIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256471.2 | XP_008254693.1 | 6 - 7 | 550 | 64 | |
| XM_008256472.2 | XP_008254694.1 | 6 - 7 | 554 | 64 | ||
| XM_008256473.2 | XP_008254695.1 | 5 - 6 | 486 | 64 | ||
More Information
Set Membership: |
|

BIN2
Genomic Map