Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265237.2 | XP_008263459.1 | 7 - 8 | 1683 | 103 | |
| XM_008265239.2 | XP_008263461.1 | 7 - 8 | 1683 | 103 | ||
| XM_008265241.2 | XP_008263463.1 | 7 - 8 | 1683 | 103 | ||
| XM_008265242.2 | XP_008263464.1 | 7 - 8 | 1682 | 103 | ||
| XM_008265243.2 | XP_008263465.1 | 7 - 8 | 1679 | 103 | ||
| XM_008265244.2 | XP_008263466.1 | 7 - 8 | 1679 | 103 | ||
| XM_008265246.2 | XP_008263468.1 | 7 - 8 | 1678 | 103 | ||
| XM_008265247.2 | XP_008263469.1 | 7 - 8 | 1687 | 103 | ||
| XM_008265248.2 | XP_008263470.1 | 7 - 8 | 1683 | 103 | ||
More Information
Set Membership: |
|

TUT4
Genomic Map