Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265237.2 | XP_008263459.1 | 9 - 10 | 1799 | 82 | |
| XM_008265239.2 | XP_008263461.1 | 9 - 10 | 1799 | 82 | ||
| XM_008265241.2 | XP_008263463.1 | 9 - 10 | 1799 | 82 | ||
| XM_008265242.2 | XP_008263464.1 | 9 - 10 | 1798 | 82 | ||
| XM_008265243.2 | XP_008263465.1 | 9 - 10 | 1795 | 82 | ||
| XM_008265244.2 | XP_008263466.1 | 9 - 10 | 1795 | 82 | ||
| XM_008265246.2 | XP_008263468.1 | 9 - 10 | 1794 | 82 | ||
| XM_008265247.2 | XP_008263469.1 | 9 - 10 | 1803 | 82 | ||
| XM_008265248.2 | XP_008263470.1 | 9 - 10 | 1799 | 82 | ||
More Information
Set Membership: |
|

TUT4
Genomic Map