Assay Details
| Gene Symbol: TMEM171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274534.2 | XP_008272756.1 | 783 | 66 | ||
| XM_008274535.2 | XP_008272757.1 | 818 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM171| Gene Symbol: TMEM171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274534.2 | XP_008272756.1 | 783 | 66 | ||
| XM_008274535.2 | XP_008272757.1 | 818 | 66 | |||
Set Membership: |
|