Assay Details
| Gene Symbol: RAB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709359.3 | XP_002709405.1 | 1 - 2 | 287 | 87 | |
| XM_008274229.2 | XP_008272451.1 | 1 - 2 | 285 | 87 | ||
| XM_017350375.1 | XP_017205864.1 | 1 - 2 | 287 | 87 | ||
More Information
Set Membership: |
|

RAB28
Genomic Map