Assay Details
| Gene Symbol: GPR156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716658.3 | XP_002716704.1 | 7 - 8 | 1428 | 65 | |
| XM_008266872.2 | XP_008265094.1 | 6 - 7 | 630 | 65 | ||
More Information
Set Membership: |
|

GPR156
Genomic Map