Assay Details
| Gene Symbol: LOC100358984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272700.2 | XP_008270922.1 | 4 - 5 | 641 | 90 | |
| XM_017349829.1 | XP_017205318.1 | 4 - 5 | 641 | 90 | ||
More Information
Set Membership: |
|

LOC100358984
Genomic Map