Assay Details
| Gene Symbol: LOC100358984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272700.2 | XP_008270922.1 | 11 - 12 | 1463 | 97 | |
| XM_017349829.1 | XP_017205318.1 | 11 - 12 | 1466 | 97 | ||
More Information
Set Membership: |
|

LOC100358984
Genomic Map