Assay Details
| Gene Symbol: LOC100358984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272700.2 | XP_008270922.1 | 13 - 14 | 1753 | 62 | |
| XM_017349829.1 | XP_017205318.1 | 13 - 14 | 1756 | 62 | ||
More Information
Set Membership: |
|

LOC100358984
Genomic Map