Assay Details
| Gene Symbol: IRAK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256751.2 | XP_008254973.1 | 11 - 12 | 1678 | 61 | |
| XM_008256753.2 | XP_008254975.1 | 10 - 11 | 1495 | 61 | ||
More Information
Set Membership: |
|

IRAK3
Genomic Map