Assay Details
| Gene Symbol: RABL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716648.3 | XP_002716694.3 | 5 - 6 | 601 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RABL3| Gene Symbol: RABL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716648.3 | XP_002716694.3 | 5 - 6 | 601 | 63 | |
Set Membership: |
|