Assay Details
| Gene Symbol: LOC100355311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267761.2 | XP_008265983.1 | 3 - 4 | 691 | 89 | |
| XM_008267762.2 | XP_008265984.1 | 3 - 4 | 703 | 89 | ||
More Information
Set Membership: |
|

LOC100355311
Genomic Map